Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABE1015

Sigma-Aldrich

Anti-monomethyl-Histone H3 (Lys79) Antibody

serum, from rabbit

Synonym(s):

Histone H3.1t, H3/t, H3t, H3/g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

yeast, human

technique(s)

ChIP: suitable (ChIP-seq)
dot blot: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

monomethylation (Lys79)

Gene Information

human ... H3C1(8350)

General description

Histone H3 is one of the five main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N-terminal tail, H3 is involved with the structure of the nucleosomes of the ′beads on a string′ structure. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine.

Immunogen

Linear peptide corresponding to human Histone H3 (Lys79).

Application

Detect Histone H3 using this rabbit polyclonal antibody, Anti-monomethyl-Histone H3 (Lys79) Antibody validated for use in western blotting, Dot Blot & ChIP-seq.
Dot Blot Analysis: Absurance Histone H3 Antibody Specificity Array (Cat. No. 16-667) which contain histone peptides with various modifications probed with Cat. No ABE1015, Anti-monomethyl-Histone H3 (Lys79) at a 1:1,000 dilution. Proteins were visualized using a Donkey anti-rabbit IgG conjugated to HRP and a chemiluminescence detection system.
Chromatin Immunoprecipitation Sequence Analysis: A representative lot used by an independent laboratory detected Histone H3 (Lys79) in NCCIT cell lysate (Jung, I., et al. (2012). Genome Res. 22(6):1026-1035).
Western Blotting Analysis: A representative lot used by an independent laboratory detected Histone H3 (Lys79) in yeast whole cell extracts (Oh. S, et al. (2010). Biochem Biophys Res Common. 399(4):512-517).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Quality

Evaluated by Western Blotting in Hela/AE cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Histone H3 (Lys79) in 10 µg of Hela/AE cell lysate.

Target description

~ 17 kDa observed

Physical form

Rabbit polyclonal in buffer containing serum with 0.05% sodium azide.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service