Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABC973

Sigma-Aldrich

Anti-BAG3

from chicken, purified by affinity chromatography

Synonym(s):

BAG family molecular chaperone regulator 3, BAG-3, Bcl-2-associated athanogene 3, Bcl-2-binding protein Bis

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

chicken

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

technique(s)

western blot: suitable

isotype

IgY

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

mouse ... Bag3(29810)

General description

BAG family molecular chaperone regulator 3 (UniProt Q9JLV1; also known as BAG-3, Bcl-2-associated athanogene 3, Bcl-2-binding protein Bis) is encoded by the Bag3 (also known as Bis, MNCb-2243) gene (Gene ID 29810) in murine species. Bcl-2-associated athanogene 3 (BAG3) is a stress-activated protein abundantly expressed in the heart, skeletal muscles, and many cancers. BAG3 is involved in protein quality control (PQC) by serving as a co-chaperone for both constitutive and inducible heat shock proteins (Hsc/Hsp), as well as in mediating cellular anti-apoptotic activity via Bcl2 binding. BAG3 also plays a role in sarcomere stability by regulating filamin clearance and production, as well as by interacting with actin capping protein beta 1 (CapZβ1) that binds the barbed end of actin and prevent actinn fibers from disssociating into actin monomers. While BAG3 is found in the cytoplasm of neonatal myocytes and involved in PQC and apoptosis, BAG3 is localized predominantly in the sarcolemma and t-tubules of adult cardiac myocytes, where it modulates myocyte contraction in response to β-adrenergic receptor signaling. Bag3-knock mice die 4 weeks after birth due to non-inflammatory myofibrillar degeneration. Human BAG3 gene mutations are found in childhood-onset muscular dystrophy, in families with dilated cardiomyopathy, as well as in cases of idiopathic dilated cardiomyopathy.

Specificity

This chicken polyclonal antibody detects mouse BAG3 by targeting a C-terminal region sequence. Rat species cross-reactivity is likely, but has not been confirmed.

Immunogen

KLH-conjugated linear peptide corresponding a sequence near the C-terminus of mouse BAG3.

Application

Detect BAG3 using this chicken polyclonal Anti-BAG3 Antibody, Cat. No. ABC973, validated for use in Western Blotting.
Research Category
Apoptosis & Cancer

Quality

Evaluated by Western Blotting in mouse heart tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected BAG3 in 10 µg of mouse heart tissue lysate.

Target description

~80 kDa observed. 61.73 kDa calculated (Met1 removed). the larger-than-calculated band size is consistent with that reported in the literature (Rasati, A., et al. (2015). Nat. Commun. 6:8695). Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity purified.
Purified chicken polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service