Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

ABC445

Sigma-Aldrich

Anti-GHRHR Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Growth hormone-releasing hormone receptor, GHRH receptor, Growth hormone-releasing factor receptor, GRF receptor, GRFR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, rat

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GHRHR(2692)

General description

GHRHR is the receptor for growth hormone release hormone (GRF, GHRF). GHRF is first produced between 18 and 29 weeks of gestation and is responsible for the production of growth hormone (GH), and GH is required for proper normal postnatal growth, bone growth, regulatory effects on protein, carbohydrate, and lipid metabolism. GHRHR is localized on cells particularly in the anterior pituitary and mutations in this gene have been associated with growth hormone deficiency, or Dwarfism of Sindh. GHRHR is localized on membranes, both plasma and nuclear membranes, and its translocation acts as a first step in the cellular signals leading to exocytotic GH secretion in the anterior pituitary.

Immunogen

KLH-conjugated linear peptide corresponding to the cytoplasmic domain of human GHRHR.

Application

Immunohistochemistry Analysis: A 1:50 dilution from a representative lot detected GHRHR in rat pituitary tissue.
This Anti-GHRHR Antibody is validated for use in Western Blotting and Immunohistochemistry (Paraffin) for the detection of GHRHR.

Quality

Evaluated by Western Blotting in human brain tissue lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected GHRHR in 10 µg of human brain tissue lysate.

Target description

~41 kDa observed. The calculated molecular weight is 47 kDa, however GHRHR has been shown as a ~41 kDa band in western blots (Zeitler, P., et al. (1998). J. Mol. Endocrinol. 21(3):363-371).

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service