ABC1437
Anti-BAG6 (BAT3)
serum, from rabbit
Synonym(s):
Large proline-rich protein BAG6, BAG family molecular chaperone regulator 6, BAG-6, BAG6, BCL2-associated athanogene 6, HLA-B-associated transcript 3, Protein G3, Protein Scythe
Select a Size
About This Item
biological source
rabbit
Quality Level
antibody form
serum
antibody product type
primary antibodies
clone
polyclonal
species reactivity
human, mouse
species reactivity (predicted by homology)
rat (based on 100% sequence homology)
technique(s)
immunocytochemistry: suitable
immunofluorescence: suitable
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
dry ice
target post-translational modification
unmodified
Gene Information
human ... BAG6(7917)
General description
Ref.:
Kawahara, H et al. (2013). J. Biochem. 153, 147-160.
Sasaki T et al. (2007). Genes Dev. 21, 848-861.
Specificity
Immunogen
Application
Immunofluorescence Analysis: A representative lot detected cytoplasmic BAG6 (Scythe) immunoreactivity in formalin-fixed paraffin-embedded human liver, pancreas, and kidney sections, while both cytoplasmic and nuclear staining was observed in one osteosarcoma biopsy tissue (Tsukahara, T., et al. (2009). Cancer Sci. 100(1):47-53).
Western Blotting Analysis: A representative lot detected both nuclear and cytoplasmic BAG6 (Scythe) localization in J774A.1 murine macrophages and bone marrow-derived macrophages. An upregulated BAG6 (Scythe) cyplasmic level and extracellular secretion was detected in both cultures upon ESAT-6 stimulation.
Western Blotting Analysis: A representative lot detected BAG6 (Scythe) in lysates from HEK293-EBNA and four human osteosarcoma cell lines, diminished target band was observed in lysates from BAG6 siRNA-transfected cells (Tsukahara, T., et al. (2009). Cancer Sci. 100(1):47-53).
Western Blotting Analysis: A representative lot detected BAG6 (Scythe) in MEF and E18.5 kidney lysates from wild-type, but not Bag6-knockout mice (Desmots, F., et al. (2005). Mol. Cell. Biol. 25(23):10329-10337).
Apoptosis & Cancer
Quality
Western Blotting Analysis: A 1:500 dilution of this antiserum detected BAG6 in 10 µg of human liver tissue lysate.
Target description
Physical form
Storage and Stability
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service