Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

ABC117

Sigma-Aldrich

Anti-NOXA1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

NADPH oxidase activator 1, NOX activator 1, Antigen NY-CO-31, NCF2-like protein, P67phox-like factor, p51-nox

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

primate (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NOXA1(10811)

General description

NOXA1 (NADPH oxidase activator 1; Antigen NY-CO-31; NCF2-like protein; P67phox-like factor; p51-nox) is an activator of the NADPH oxidase NOX1. It contains multiple domains, including four TPR repeats which facilitate interaction with RAC1, and one SH3 domain that binds to NOXO1. NOXA1 interacts with NOXO1, RAC1, NOX1, and CYBA to form a multi-protein complex that produces reactive oxygen species for important cellular processes such as oxygen sensing, hormone biosynthesis, and signal transduction. NOXA1 is found in pancreas, spleen, liver, prostate, colon, and small intestine.

Specificity

No homology to Rat or Mouse detected.

Immunogen

KLH-conjugated linear peptide corresponding to human NOXA1.

Application

Detect NOXA1 using this Anti-NOXA1 Antibody validated for use in Western Blotting.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Quality

Evaluated by Western Blot in Human Colon tissue lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected NOXA1 in 10 µg of human colon tissue lysate.

Target description

~51 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human colon tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service