Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB9838

Sigma-Aldrich

Anti-Rabphillin 3A Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

Specificity

Rabphilin 3A. The antibody recognizes a protein of ~82 kDa corresponding to Rabphilin 3A in lysates from rat cortex.
The immunogen has 100% homology with human, non-human primates, mouse, bovine and canine.

Immunogen

Synthetic peptide from the C-terminal region of rat Rabphilin 3A.

Application

This Anti-Rabphillin 3A Antibody is validated for use in WB for the detection of Rabphillin 3A.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service