Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB9754

Sigma-Aldrich

Anti-Kisspeptin Antibody

serum, Chemicon®

Synonym(s):

kp10

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

sheep, rat, mouse

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable
radioimmunoassay: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Specificity

Kisspeptin 10 (kp10, metastin). No cross reactivity with GnRH, Galatin, SP, NPY, CRH, alphaMSH, SRIF or PrRP by RIA.
The antibody showed a very low level of reactivity with human kp10.

Immunogen

Peptide from mouse kisspeptin 10. The sequence is common to all kisspeptin isoforms.

Application

Anti-Kisspeptin Antibody detects level of Kisspeptin & has been published & validated for use in IH, RIA.
Immunohistochemistry on 4% paraformaldehyde fixed tissue: 1:500-1:1,000 (PAP and ABC with NiDAB detection system).

RIA: 1:500-1:10,000

Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Hormones & Receptors

CNS Control of Metabolism

Physical form

Rabbit serum.Liquid containing 50% glycerol, Ph 7.4, Phosphate buffer 0.01M.
Serum

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R M Cravo et al.
Neuroscience, 173, 37-56 (2010-11-26)
Humans and mice with loss-of-function mutations of the genes encoding kisspeptins (Kiss1) or kisspeptin receptor (Kiss1r) are infertile due to hypogonadotropic hypogonadism. Within the hypothalamus, Kiss1 mRNA is expressed in the anteroventral periventricular nucleus (AVPV) and the arcuate nucleus (Arc).
Leptin deficiency and diet-induced obesity reduce hypothalamic kisspeptin expression in mice.
Quennell, JH; Howell, CS; Roa, J; Augustine, RA; Grattan, DR; Anderson, GM
Endocrinology null
Kisspeptin immunoreactive cells of the ovine preoptic area and arcuate nucleus co-express estrogen receptor alpha.
Franceschini, I, et al.
Neuroscience Letters, 401, 225-230 (2006)
Characterisation of Arcuate Nucleus KisspeptinNeurokinin B Neuronal Projections and Regulation during Lactation in the Rat.
True C
Journal of Neuroendocrinology null
Anabolic androgenic steroid abuse: multiple mechanisms of regulation of GABAergic synapses in neuroendocrine control regions of the rodent forebrain.
J G Oberlander,D M Porter,C A A Penatti,L P Henderson
Journal of Neuroendocrinology null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service