Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB9479

Sigma-Aldrich

Anti-Putative Neurotransmitter Receptor Antibody

Chemicon®, from rabbit

Synonym(s):

PNR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable (paraffin)

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... TAAR5(9038)

Specificity

Recognizes Putative Neurotransmitter Receptor [PNR].

Immunogen

Synthetic peptide from the 2nd extracellular domain of human Putative Neurotransmitter Receptor.

Application

Detect Putative Neurotransmitter Receptor using this Anti-Putative Neurotransmitter Receptor Antibody validated for use in IH(P).

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service