Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB9313

Sigma-Aldrich

Anti-Vasopressin V2 Receptor. Antibody

Chemicon®, from rabbit

Synonym(s):

V2 Receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable (paraffin)

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... AVPR2(554)

Specificity

Other species have not been tested. The immunogen has 67% homology with mouse and 67% with rat.
Recognizes Vasopressin V2 Receptor.

Immunogen

Synthetic peptide from the C-terminal domain of human Vasopressin V2 Receptor.

Application

Detect Vasopressin V2 Receptor. using this Anti-Vasopressin V2 Receptor. Antibody validated for use in IH(P).
Immunohistochemistry on paraffin embedded tissue sections.
Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Neurochemistry & Neurotrophins

Neuroinflammation & Pain

Physical form

Liquid in PBS, pH 7.7 with 0.01% sodium azide.

Storage and Stability

Maintain at -70°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service