Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB5846

Sigma-Aldrich

Anti-GABA A Receptor Associated Protein Antibody

Chemicon®, from rabbit

Synonym(s):

GABARAP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat, mouse, human

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... GABARAP(11337)

General description

GABAA Receptor Associated Protein (GABARAP) interacts with the gamma2 subunit of GABAA Receptors

Specificity

GABA-A Receptor Associated Protein (GABARAP), N-terminus.

Immunogen

Synthetic peptide from human GABARAP.

Application

Immunoblotting: 1:500-1:1,000. Recognizes a protein of ~14 kDa.

Optimal working dilutions must be determined by end user.
This Anti-GABA A Receptor Associated Protein Antibody is validated for use in WB for the detection of GABA A Receptor Associated Protein.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service