Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB3245-I-AF488

Sigma-Aldrich

Anti-TopBP1 Antibody, Alexa Fluor 488 Conjugate

from rabbit, ALEXA FLUOR 488

Synonym(s):

DNA topoisomerase 2-binding protein 1, DNA topoisomerase II-beta-binding protein 1, DNA topoisomerase II-binding protein 1, TopBP1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

biological source

rabbit

conjugate

ALEXA FLUOR 488

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... TOPBP1(11073)

General description

DNA topoisomerase 2-binding protein 1 (UniProt Q92547; also known as DNA topoisomerase II-beta-binding protein 1, DNA topoisomerase II-binding protein 1, TopBP1) is encoded by the TOPBP1 (also known as KIAA0259) gene (Gene ID 11073) in human. Timely DNA replication and accurate chromosome segregation is critical for maintaining genome integrity to avoid cancer and other genetic diseases. TopBP1 plays an important role in mediating DNA processing during mitosis to reduce transmission of DNA damage toG1 daughter cells. Unlike the translesion synthesis scaffold factor Rev1 or K164-ubiquitylated PCNA that serve to recruit translesion polymerases to stalled forks, TopBP1 forms foci upon mitotic entry, marking sites of and promoting unscheduled DNA synthesis at underreplicated regions independent of Rev1 and PCNA. Persistent TopBP1 foci transition into 53BP1 nuclear bodies (NBs) in G1 and precise temporal depletion of TopBP1 just before mitotic entry induces formation of 53BP1 NBs in the next cell cycle. Human TopBP1 contains eight BRCA1 C Terminus (BRCT) domains (a.a. 101-189, 195-284, 354-444, 548-633, 641-738, 900-991, 1259-1351, 1389-1486) and two nuclear localization signal (NLS) motifs (a.a. 852-858, 1517-1520). TopBP1 BRCT1/2-mediated interaction with phosphorylated Rad9 is essential for TopBP1-dependent ATR (ataxia telangiectasia and Rad3-related) activation, while TopBP1 BRCT6 interacts with the phosphorylated transcription factor, E2F1 to repress E2F1 activity in response to DNA damage. BRCT6 is also targeted for poly(ADP-ribosyl)ation by poly(ADP-ribose) polymerase 1 (PARP-1). The innate immune regulator STAT-5 is activated by human papillomavirus (HPV) proteins in HIV-infected cells, resulting in upregulated TopBP1 transcription and ATR pathway activation required for HPV genome amplification.

Specificity

This rabbit polyclonal antibody targets human TopBP1 region between the sixth and seventh BRCA1 C Terminus (BRCT) domain.

Immunogen

KLH-conjugated linear peptide corresponding to a squence from the region between BRCT domain 6 and 7 of human TopBP1.

Application

Anti-TopBP1, Alexa Fluor 488 Conjugate, Cat. No. AB3245-I-AF488, is a highly specific rabbit polyclonal antibody that targets and has been tested in Immunocytochemistry.
Research Category
Epigenetics & Nuclear Function
The unconjugated antibody (Cat. No. AB3245) is also available for Immunoprecipitation and Western Blotting applications.

Quality

Evaluated by Immunocytochemistry in HeLa cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody conjugate detected TopBP1 in 4% paraformaldehyde-fixed, 0.1% Triton X-100-permeabilized HeLa cells.

Target description

~180 kDa observed. 170.7 kDa calculated.

Physical form

Affinity purified.
Purified rabbit polyclonal antibody conjugate in PBS with 1.5% BSA and 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service