Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB2915

Sigma-Aldrich

Anti-Bax Antibody

serum, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... BAX(581)

General description

The previously assigned protein identifier Q07814 has been merged into Q07812. Full details can be found on the UniProt database.

Specificity

Recognizes Mouse and Rat Bax-alpha, a 21 kDa protein, as well as a longer 24 kDa. Bax-beta isoform and a 5 kDa Bax-gamma isoform, which are present in much smaller amounts in most tissues. Does not react with other members of the Bcl-2 family, as verified by immunoblot.

Immunogen

A synthetic peptide corresponding to amino acids 43-61 of the mouse Bax-alpha protein.

Application

Immunohistochemistry (ABC method) on formalin-fixed paraffin-embedded sections (1:600-1:1500)

Immunoblot (1;2500) using ECL detection.

High levels of Bax-alpha are found in areas of the brain, including the cerebrum and cerebellum, as well as the small intestine, stomach, overy, testis, uterus, thymus, and spleen.

Optimal working dilutions must be determined by end user.
Research Category
Apoptosis & Cancer
Research Sub Category
BCL2 & Inhibition
This Anti-Bax Antibody is validated for use in WB, IH(P) for the detection of Bax.

Linkage

Replaces: 04-434

Physical form

Liquid antiserum, no preservatives.

Storage and Stability

Store at -20°C for up to 12 months in undiluted aliquots. Avoid repeated freeze-thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Malekzadeh Yasin et al.
Environmental toxicology, 33(11), 1204-1213 (2018-08-21)
The present experimental study was carried out to investigate the crosslink between aflatoxin B1 (AFB1)-induced oxidative stress and mitochondria-dependent apoptosis in testicles. For this purpose, 24 mature male Swiss albino mice were randomly divided into control and test groups. The
Mohammad Ali Maadi et al.
Andrologia, 54(3), e14349-e14349 (2021-12-10)
Testicular torsion (TT) is an emergency complication that leads to oxidative stress and adversely affects spermatogenesis. Although immediate treatment consists of testicular detorsion (TD) to reverse TT-induced ischemia, mechanisms underlying recovery have yet to be fully understood. The current study
Hojat Anbara et al.
Environmental toxicology, 36(2), 223-237 (2020-09-21)
Aspartame is one of the most common consumed artificial sweeteners utilized in many food products and beverages. It has been indicated that long-term consumption of aspartame leads to reproductive toxicity but its mechanism is not well-clear. In this study we
Barkha P Patel et al.
PloS one, 5(2), e9386-e9386 (2010-03-03)
Caloric restriction (CR) extends lifespan through a reduction in oxidative stress, delays the onset of morbidity and prolongs lifespan. We previously reported that long-term CR hastened clinical onset, disease progression and shortened lifespan, while transiently improving motor performance in G93A
Ying-Yan Fang et al.
Journal of neurochemistry, 146(6), 703-721 (2018-06-26)
Menopause, a risk factor for brain dysfunction in women, is characterized by neuropsychological symptoms including depression and dementia, which are closely related to alterations in different brain regions after menopause. However, little is known about the variability in pathophysiologic changes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service