Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB2212

Sigma-Aldrich

Anti-phospho GluR4 (S862) Antibody

serum, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer862)

Gene Information

human ... GRIA4(2893)

General description

Glutamate receptors are the predominant excitatory neurotransmitter receptors in the mammalian brain and are activated in a variety of normal neurophysiologic processes. These receptors are heteromeric protein complexes composed of multiple subunits, arranged to form ligand-gated ion channels. The classification of glutamate receptors is based on their activation by different pharmacologic agonists. Alternative splicing of this gene results in transcript variants encoding different isoforms, which may vary in their signal transduction properties. Some haplotypes of this gene show a positive association with schizophrenia. Direct phosphorylation of GluR4-containing receptors may regulate rapid excitatory synaptic transmission. In addition, phosphorylation of GluR4 S862. is both necessary and sufficient to deliver the receptor to the synapse and induce plasticity in early postnatal development.

Specificity

Cat. # AB2212 recognzies phospho GluR4 (S862).
May react with mouse, human, primate and dog based on sequence homology. Reactivity with other species has not been tested.

Immunogen

Synthetic peptide from rat.

Application

Anti-phospho GluR4 (S862) Antibody is an antibody against phospho GluR4 (S862) for use in WB.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors
Western Blot Analysis:
Cat. # AB2212 recognizes pGluR4 (S862) at 1:5,000 in rat brain cerebellum Lysate.

Quality

Routinely tested on rat brain cerebellum lysate.

Target description

~105 kDa

Physical form

Serum with 0.05% NaN3

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 1 year after date of receipt.

Analysis Note

Control
Rat Brain Cerebellum lysate

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pain after discontinuation of morphine treatment is associated with synaptic increase of GluA4-containing AMPAR in the dorsal horn of the spinal cord.
Caba?ero, D; Baker, A; Zhou, S; Hargett, GL; Irie, T; Xia, Y; Beaudry, H; Gendron et al.
Neuropsychopharmacology null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service