Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB2039

Sigma-Aldrich

Anti-Elastin Antibody

Chemicon®, from rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
radioimmunoassay: suitable

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... ELN(2006)

Specificity

Antibody shows less than 0.1% reactivity with rat Collagen Types I, II, III, V and human Elastin by radioimmunoassay.

Immunogen

Elastin extracted and purified from rat aorta.

Application

Immunohistochemistry: 1:40 dilution for immunofluorescent staining of frozen rat skin and liver tissues.

Radioimmunoassay

ELISA: 1:100-1:200

Optimal working dilutions must be determined by end user.
Research Category
Cell Structure
Research Sub Category
Integrins
This Anti-Elastin Antibody is validated for use in ELISA, RIA, IH for the detection of Elastin.

Physical form

Affinity purified immunoglobulin. Liquid in 0.01M phosphate, 0.09M NaCl, pH7.2 with no preservatives.

Storage and Stability

Maintain frozen at -20°C for up to 12 months. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Matthias Roth-Kleiner et al.
Histochemistry and cell biology, 141(1), 75-84 (2013-08-06)
Pre- and postnatal corticosteroids are often used in perinatal medicine to improve pulmonary function in preterm infants. To mimic this clinical situation, newborn rats were treated systemically with dexamethasone (Dex), 0.1-0.01 mg/kg/day on days P1-P4. We hypothesized that postnatal Dex may
Jonah D Lee et al.
Clinical oral investigations, 26(7), 5019-5027 (2022-04-02)
The aim of this study was to compare and characterize the structural and ultrastructural organization of the temporomandibular joint (TMJ) between two large animal models for use in the development of tissue engineering strategies. Whole TMJs from sheep and pigs
A A Phillips et al.
The Journal of physiology, 594(6), 1677-1688 (2015-12-05)
High-thoracic or cervical spinal cord injury (SCI) is associated with several critical clinical conditions related to impaired cerebrovascular health, including: 300-400% increased risk of stroke, cognitive decline and diminished cerebral blood flow regulation. The purpose of this study was to
Aaron A Phillips et al.
Journal of neurotrauma, 35(3), 573-581 (2017-11-17)
We aimed to create a clinically relevant pre-clinical model of transient hypertension, and then evaluate the pathophysiological cerebrovascular processes resulting from this novel stimulus, which has recently been epidemiologically linked to cerebrovascular disease. We first developed a clinically relevant model
Spatial and temporal analysis of extracellular matrix proteins in the developing murine heart: a blueprint for regeneration.
Hanson, KP; Jung, JP; Tran, QA; Hsu, SP; Iida, R; Ajeti, V; Campagnola, PJ; Eliceiri et al.
Tissue Engineering: Part A null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service