Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB15542

Sigma-Aldrich

Anti-Myelin Basic Protein Antibody

CHEMICON®, rabbit polyclonal

Synonym(s):

MBP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Product Name

Anti-Myelin Basic protein Antibody, serum, Chemicon®

biological source

rabbit

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

packaging

antibody small pack of 25 μL

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MBP(4155)

Specificity

Myelin basic protein [MBP]. By Western blot the antibody recognizes a band at ~14-18 kDa on human brain lysate and ~14-18 kDa on recombinant MBP. An additional band at ~65 kDa may be seen depending on sample and antibody concentration used.

Immunogen

Recombinant MBP.

Application

Detect Myelin Basic protein using this Anti-Myelin Basic protein Antibody validated for use in WB.
Research Category
Neuroscience
Research Sub Category
Neuronal & Glial Markers

Neurochemistry & Neurotrophins
Western blot: 1:1,000-1:2,000 using recombinant MBP and 1:5,000-1:10,000 using human brain lysate.

Optimal working dilutions must be determined by end user.

Quality

Tested

Physical form

Liquid.
Serum

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Other Notes

Please refer to lot specific datasheet.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vidhu Sharma et al.
eLife, 5 (2016-02-20)
The Reproducibility Project: Cancer Biology seeks to address growing concerns about reproducibility in scientific research by conducting replications of selected experiments from a number of high-profile papers in the field of cancer biology. The papers, which were published between 2010

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service