Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB15396

Sigma-Aldrich

Anti-acetyl BMAL1 (Lys538) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

ARNT-like protein 1, brain and muscle, Basic-helix-loop-helix-PAS protein MOP3, Brain and muscle ARNT-like 1, Member of PAS protein 3, aryl hydrocarbon receptor nuclear translocator-like, bHLH-PAS protein JAP3, basic-helix-loop-helix-PAS orphan MOP3, mem

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity (predicted by homology)

primate, opossum, mouse, rat, pig, horse

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

Gene Information

General description

Brain and muscle Arnt-like protein-1 (BMAL1; also known as MOP3 or Arnt3) is a transcription factor known to regulate circadian rhythm. BMAL1 is the only component of the mammalian circadian clock whose sole deletion in a mouse model generates arrhythmicity. In addition to defects in the clock, these BMAL1 null-mice also have reproductive problems are small in stature, age quickly and have progressive arthropathy that results in having less overall locomotor activity than wild type mice. Recent phenotyping data suggests that BMAL1 and its partner Clock also play a role in regulation of glucose homeostasis and metabolism. Finally, BMAL1, NPAS2, and PER2 have been associated with seasonal affective disorder in humans. BMAL1 transcription is circadian and reciprocally regulated by NR1D1 (Rev-erb-alpha) and RORA, which establishes a second interlocking loop in the mammalian circadian clock.

Specificity

Cat. No. AB15396 recognizes acetylated lysine (Kac) within the –GG[Kac]KI LN— sequence present in BMAL1. The (Lys538) designation is based on human isoform BMAL1B (Uniprot O00327-2; also known as JAP3). The lysine numbering varies from spliced isoform to isoform, and from species to species. Note: Lys 537 mentioned in the publication by Hirayama, J. et al. (2007). Nature. 450(7172):1086-1090, in fact corresponds to Lys538 based on the Uniprot sequence O00327-2

Immunogen

Epitope: C-Terminus
KLH-conjugated BMAL1 acetylated at Lysine538.

Application

Anti-acetyl BMAL1 (Lys538) Antibody is an antibody against acetyl BMAL1 (Lys538) for use in WB.
Research Category
Neuroscience
Research Sub Category
Circadian Rhythm & Sleep

Target description

Approx. 95 kDa

Physical form

Antigen Affinity Purified
Antigen Affinity purified Rabbit polyclonal, supplied at 1 mg/mL, containing 0.1M Tris-Glycine (pH7.4) 150mM NaCl with 0.05% NaN3.

Storage and Stability

Stable for 1 year at 2-8ºC from date of receipt.

Analysis Note

Control
Transfected HEK293/myc-BMAL1/empty
vector and HEK293/myc-BMAL1/myc-CLOCK
treated with Nicotinamide.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10-13 - German Storage Class 10 to 13


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nikolai Petkau et al.
eLife, 8 (2019-07-12)
Many physiological processes exhibit circadian rhythms driven by cellular clocks composed of interlinked activating and repressing elements. To investigate temporal regulation in this molecular oscillator, we combined mouse genetic approaches and analyses of interactions of key circadian proteins with each
Kevin B Koronowski et al.
Cell, 177(6), 1448-1462 (2019-06-01)
Mammals rely on a network of circadian clocks to control daily systemic metabolism and physiology. The central pacemaker in the suprachiasmatic nucleus (SCN) is considered hierarchically dominant over peripheral clocks, whose degree of independence, or tissue-level autonomy, has never been
Shogo Sato et al.
Cell, 170(4), 664-677 (2017-08-13)
The process of aging and circadian rhythms are intimately intertwined, but how peripheral clocks involved in metabolic homeostasis contribute to aging remains unknown. Importantly, caloric restriction (CR) extends lifespan in several organisms and rewires circadian metabolism. Using young versus old
Kenichiro Kinouchi et al.
Cell reports, 25(12), 3299-3314 (2018-12-20)
The circadian clock operates as intrinsic time-keeping machinery to preserve homeostasis in response to the changing environment. While food is a known zeitgeber for clocks in peripheral tissues, it remains unclear how lack of food influences clock function. We demonstrate
Daniel C Levine et al.
Molecular cell, 78(5), 835-849 (2020-05-06)
Disrupted sleep-wake and molecular circadian rhythms are a feature of aging associated with metabolic disease and reduced levels of NAD+, yet whether changes in nucleotide metabolism control circadian behavioral and genomic rhythms remains unknown. Here, we reveal that supplementation with

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service