Nicotinic acetylcholine receptors, also known as nAChRs, belong to a family of proteins called ligand gated ion channels and are widely expressed throughout the nervous system. nAChRs are made up of 5 subunits (two α and three β). Subunits α4 and β2 (α4β2) is the main receptor in the mammalian brain and has been associated with the positive reinforcing effects of nicotine. The relative contribution of nAChRs β2 and β4 are not well understood because the subunit structure that mediates synaptic transmission in the peripheral nervous system is unknown.
Specificity
This antibody recognizes Nicotinic Acetylcholine Receptor β2.
Immunogen
This antibody is generated from rabbits immunized with a KLH-conjugated linear peptide selected from residues of Nicotinic Acetylcholine Receptor β2.
Application
Detect Nicotinic Acetylcholine Receptor β2 using this Anti-Nicotinic Acetylcholine Receptor β2 Antibody validated for use in WB.
Quality
Routinely evaluated by Western Blot in rat e16 brain lysate. Western Blot Analysis: : 1:20,000 dilution of this lot detected Nicotinic Acetylcholine Receptor β2 on 10 µg of rat e16 brain lysate.
Target description
Approx. 55 kDa
Other Notes
Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 37(47), 11377-11389 (2017-10-25)
Acetylcholine (ACh) is a potent neuromodulator capable of modifying patterns of acoustic information flow. In auditory cortex, cholinergic systems have been shown to increase salience/gain while suppressing extraneous information. However, the mechanism by which cholinergic circuits shape signal processing in
The thalamocortical pathways form highly topographic connections from the primary sensory thalamic nuclei to the primary cortical areas. The synaptic properties of these thalamocortical connections are modifiable by activation from various neuromodulators, such as acetylcholine. Cholinergic activation can alter functional
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.