AB15134
Anti-Neurotensin Receptor 2 Antibody
from rabbit, purified by affinity chromatography
Synonym(s):
Levocabastine-sensitive neurotensin receptor, NTR2 receptor, neurotensin receptor 2, neurotensin receptor, type 2
About This Item
Recommended Products
biological source
rabbit
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
human, mouse, rat
technique(s)
immunohistochemistry: suitable
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
Gene Information
human ... NTSR2(23620)
General description
Specificity
Immunogen
Application
Neuroscience
Metabolism
Neurotransmitters & Receptors
Hormones & Receptors
Western Blot Analysis: 1 µg/ml of this antibody detected Neurotensin Receptor 2 in 10 µg of rat brain lysate.
Quality
Western Blot Analysis: 1 µg/ml of this antibody detected Neurotensin Receptor 2 in 10 µg of human cortex lysate.
Target description
Linkage
Physical form
Storage and Stability
Analysis Note
Human cortex lysate
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service