Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB1413-I-AF488

Sigma-Aldrich

Anti-Interleukin-1β Antibody, Alexa Fluor 488 Conjugate

from rabbit, ALEXA FLUOR 488

Synonym(s):

Interleukin-1 beta, IL-1 beta, IL-1b, IL-1beta

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$401.00

$401.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$401.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

$401.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

conjugate

ALEXA FLUOR 488

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse

species reactivity (predicted by homology)

gerbil (based on 100% sequence homology), rat (based on 100% sequence homology), hamster (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

gerbil ... Il1B(110551400)
hamster ... Il1B(100775079)
mouse ... Il1B(16176)
rat ... Il1B(24494)

General description

Interleukin-1 beta (UniProt; also known as IL-1 beta, IL-1b, IL-1beta) is encoded by the Il1b gene (Gene ID 16176) in murine species. Interleukin-1 (IL-1) is a cytokine that plays a central role in the regulation of acute and chronic inflammation. Two forms of IL-1, IL-1 and IL-1 , are transcribed from two separate genes and mediate similar, but not completely overlapping, biological effects via the membrane-bound IL-1 type 1 receptor (IL-1R1). Studies utilizing Il1a- and Il1b-knockout mice showed that IL-1 , but not IL-1 , is involved in T-cell-dependent antibody production via antigen-specific T-cell activation. IL-1 binding to the extracellular domain of IL1R1 promotes the recruitment of the IL-1 receptor accessory protein and subsequent signal transduction mediated by the cytoplasmic domains of IL-1R1 and IL1-R2. The IL-1 type 2 receptor (IL1R2) is unable to initiate signaling and only acts as a decoy receptor. Soluble receptors for IL-1 (sIL-1Rs) are produced by proteolytic cleavage of membrane-bound receptors or by alternative splicing and act as natural inhibitors by competing against membrane-bound IL1Rs for IL-1 binding. Murine IL-1 beta is produced with a propeptide sequence (a.a. 1-117), which is removed posttranslationally to yield the mature (a.a. 118-269) protein.

Specificity

The unconjugated antibody (Cat. No. AB1413-I) detected both the ~17 kDa mature and ~30 kDa pro-form of murine IL-1beta. The corresponding sequence in human IL-1beta differs from that of the immunogen by only one amino acid. Immunogen sequence is not found in IL-1alpha.

Immunogen

KLH-conjugated linear peptide corresponding to a sequence within the C-terminal half of murine and rat mature IL-1beta.

Application

Research Category
Inflammation & Immunology
The unconjugated antibody (Cat. No. AB1413-I) and Alexa Fluor 555 conjugate (Cat. No. AB1413-I-AF555) are also available for Immunocytochemistry, Immunoprecipitation, and Western Blotting applications.
This rabbit polyclonal Anti-Interleukin-1 Antibody, Alexa Fluor 488 Conjugate, Cat. No. AB1413-I-AF488, is validated for use in Immunocytochemistry for the detection of Interleukin-1 .

Quality

Evaluated by Immunocytochemistry in NIH/3T3 cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected IL-1beta in 4% paraformaldehyde-fixed, Triton X-100-permeabilized NIH/3T3 cells.

Target description

~17/30 kDa observed. 17.39/30.93 kDa (mouse mature/pro-form), 17.32/30.64 kDa (rat mature/pro-form) calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Affinity purified.
Purified rabbit polyclonal antibody conjugate in PBS with 1.5% BSA and 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service