Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB10422

Sigma-Aldrich

Anti-Phospho-Bad (Ser134) Antibody

from rabbit

Synonym(s):

BCL-X/BCL-2 binding protein, BCL2-antagonist of cell death protein, BCL2-associated agonist of cell death, BCL2-binding component 6, BCL2-binding protein, Bcl-2-binding component 6, Bcl-2-like 8 protein, Bcl-XL/Bcl-2-associated death promoter

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

purified antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer134)

Gene Information

human ... BAD(572)

General description

Bad is a member of the BCL-2 family. BCL-2 family members are known to be regulators of programmed cell death. This protein positively regulates cell apoptosis by forming heterodimers with BCL-xL and BCL-2, and reversing their death repressor activity. Proapoptotic activity of this protein is regulated through its phosphorylation. Protein kinases AKT and MAP kinase, as well as protein phosphatase calcineurin are found to be involved in the regulation of this protein. Bad is phosphorylated on one or more of Ser-75, Ser-99, Ser-118 and Ser-134 in response to survival stimuli, which blocks its pro-apoptotic activity. Phosphorylation on Ser-99 or Ser-75 promotes heterodimerization with 14-3-3 proteins. This interaction then facilitates the phosphorylation at Ser-118, a site within the BH3 motif, leading to the release of Bcl-X(L) and the promotion of cell survival. Ser-99 is the major site of AKT/PKB phosphorylation, Ser-118 the major site of protein kinase A (CAPK) phosphorylation.

Specificity

Other species have not been tested.
The antibody reacts with the 18 kDa Phospho-Bad-S134 domain protein.

Immunogen

This antibody is generated from rabbits immunized with a KLH conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding S134 of human Bad.

Application

Anti-Phospho-Bad (Ser134) Antibody detects level of Phospho-Bad (Ser134) & has been published & validated for use in WB.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Quality

Western Blot Analysis: 1:250 dilution of this antibody detected Phospho-Bad-S134 on Jurkat lysate.

Target description

18 kDa

Physical form

Format: Purified
Protein G followed by IAP
Purified in PBS with 0.09% NaN3.

Storage and Stability

Store at 2-8°C for 6 months from date of receipt. For long term storage, store at -20°C.

Analysis Note

Control
Western Blot : Jurkat lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service