Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

AB10324

Sigma-Aldrich

Anti-CYP450 Pan 2C Antibody

serum, Chemicon®

Synonym(s):

cytochrome P450

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
Product AB10324 is not currently sold in your country. Contact Technical Service

biological source

rabbit

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CYP2C8(1558)

Immunogen

Recombinant CYP450 2C19

Application

Research Category
Metabolism
Research Sub Category
Enzymes & Biochemistry
This Anti-CYP450 Pan 2C Antibody is validated for use in WB for the detection of CYP450 Pan 2C.

Quality

Routinely evaluated by immunoblot.

Target description

56 kDa

Physical form

Raw Rabbit Serum with 0.1% Sodium Azide
Serum

Storage and Stability

1 year at 2-8°C from date of shipment

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service