8.52241
Fmoc-D-Thr(Trt)-OH
Novabiochem®
Synonym(s):
Fmoc-D-Thr(Trt)-OH, N-α-Fmoc-O-trityl-D-threonine / (2R,3S)
Select a Size
About This Item
Recommended Products
Quality Level
product line
Novabiochem®
assay
≥78.0% (acidimetric)
≥97.0% (HPLC)
≥98% (TLC)
form
powder
reaction suitability
reaction type: Fmoc solid-phase peptide synthesis
manufacturer/tradename
Novabiochem®
application(s)
peptide synthesis
functional group
hydroxyl
Storage temp.
15-25°C
SMILES string
N([C@H]([C@@H](OC(c6ccccc6)(c5ccccc5)c4ccccc4)C)C(=O)O)C(=O)OCC1c2c(cccc2)c3c1cccc3
Inchi Key
JARBLLDDSTVWSM-OSPAZUARSA-N
General description
Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS
Guide to Selection of Orthogonally-Protected Building Blocks for Fmoc SPPS
Linkage
Analysis Note
Appearance of substance (visual): powder
Identity (IR): passes test
Enantiomeric purity: ≥ 99.0 % (a/a)
Purity (TLC(011A)): ≥ 98 %
Purity (TLC(0811)): ≥ 98 %
Assay (HPLC, area%): ≥ 97.0 % (a/a)
Solubility (1 mmole in 2 ml DMF): clearly soluble
Assay (acidimetric): ≥ 78.0 %
Water (K. F.): ≤ 1.5 %
To see the solvent systems used for TLC of Novabiochem® products please click here.
Legal Information
Storage Class
11 - Combustible Solids
wgk_germany
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Novabiochem® offers orthogonally protected amino acids for peptide synthesis, including cyclic and branched peptides.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service