8.52223
Fmoc-Ile-OPfp
Novabiochem®
Synonym(s):
Fmoc-Ile-OPfp, N-α-Fmoc-L-isoleucine pentafluorophenyl ester
About This Item
Quality Level
product line
Novabiochem®
assay
≥98.0% (HPLC)
form
powder
reaction suitability
reaction type: Fmoc solid-phase peptide synthesis
manufacturer/tradename
Novabiochem®
application(s)
peptide synthesis
functional group
Fmoc
storage temp.
15-25°C
InChI
1S/C27H22F5NO4/c1-3-13(2)24(26(34)37-25-22(31)20(29)19(28)21(30)23(25)32)33-27(35)36-12-18-16-10-6-4-8-14(16)15-9-5-7-11-17(15)18/h4-11,13,18,24H,3,12H2,1-2H3,(H,33,35)/t13-,24-/m0/s1
InChI key
FYZSBHSHLNJGPS-RZFZLAGVSA-N
Related Categories
General description
Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS
Linkage
Analysis Note
Appearance of substance (visual): powder
Identity (IR): passes test
Optical rotation α 25/D (c=1 in chloroform): -15.0 - -12.0 °
DCU (DC (0004)): ≤ 5 %
Assay (HPLC, area%): ≥ 98.0 % (a/a)
Solubility (0,5 mmole in 3 ml DMF): clearly soluble
To see the solvent systems used for TLC of Novabiochem® products please click here.
Legal Information
signalword
Warning
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service