Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.45157

Sigma-Aldrich

1,1,1,3,3,3-Hexafluoro-2-propanol

Msynthplus

Synonym(s):

1,1,1,3,3,3-Hexafluoro-2-propanol, HFP, Hexafluoroisopropanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3CH(OH)CF3
CAS Number:
Molecular Weight:
168.04
MDL number:
UNSPSC Code:
12352104
EC Index Number:
213-059-4

vapor pressure

269 hPa ( 30 °C)

Quality Level

assay

≥99.0% (GC)

form

liquid

potency

1500 mg/kg LD50, oral (Rat)

pH

3-4 ( in H2O, Aqueous solution)

bp

58 °C/1013 hPa

mp

-3 °C

transition temp

flash point >100 °C

density

1.62 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C3H2F6O/c4-2(5,6)1(10)3(7,8)9/h1,10H

InChI key

BYEAHWXPCBROCE-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Colour number (Hazen): ≤ 20
Density (d 20 °C/ 4 °C): 1.614 - 1.618
Identity (IR): passes test
Pd (Palladium) (ICP MS): lot specific result

Legal Information

MSYNTH is a trademark of Merck KGaA, Darmstadt, Germany

pictograms

CorrosionHealth hazard

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Repr. 2 - Skin Corr. 1A - STOT RE 2

Storage Class

8A - Combustible, corrosive hazardous materials

wgk_germany

WGK 2

flash_point_c

closed cup - boils before flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service