Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.45151

Sigma-Aldrich

Tetramethylammonium hydroxide

(25% solution in water) Msynthplus

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352107

vapor pressure

23.33 hPa ( 20 °C)

pH

>13 (20 °C in H2O)

bp

110 °C/1013 hPa

density

1.02 g/cm3 at 20 °C

storage temp.

2-30°C

Analysis Note

Assay (acidimetric): 24.0 - 27.0 %
Color number (Hazen): ≤ 50
The analysis of residual solvents is not possible due to the physicochemical properties of the material.

Legal Information

MSYNTH is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service