Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.45111

Sigma-Aldrich

Piperidine

Msynthplus

Synonym(s):

Piperidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H11N
CAS Number:
Molecular Weight:
85.15
MDL number:
UNSPSC Code:
12352005
EC Index Number:
203-813-0

vapor pressure

34 hPa ( 20 °C)

assay

≥99% (GC)

form

liquid

autoignition temp.

320 °C

potency

276 mg/kg LD50, skin (Rabbit)

expl. lim.

1.5-10.3 % (v/v)

pH

12.6 (20 °C, 100 g/L in H2O)

kinematic viscosity

1.77 cSt(20 °C)

bp

106 °C/1013 hPa

mp

-10.8 °C

transition temp

flash point 16 °C

density

0.86 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C5H11N/c1-2-4-6-5-3-1/h6H,1-5H2

InChI key

NQRYJNQNLNOLGT-UHFFFAOYSA-N

General description

Msynthplus

Application

Piperidine Msynthplus. CAS 110-89-4, pH 12.6 (100 g/l, H₂O, 20 °C).

Analysis Note

Appearance of substance (visual)clearColour number (Hazen)≤ 50Assay (GC, area%)≥ 99.0 % (a/a)Density (d 20 °C/ 4 °C)0.861 - 0.863Water (K. F.)≤ 0.50 %Identity (IR)passes test1,1-Dichloroethene (HS-GC)lot specific result1,1,1-Trichloroethane (HS-GC)lot specific resultBenzene (HS-GC)lot specific resultCarbon tetrachloridenot measurable1,2-Dichlorethanenot measurable

Legal Information

MSYNTH is a trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_c

16 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service