Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.45105

Sigma-Aldrich

Tetramethylammonium chloride

Msynthplus

Synonym(s):

Tetramethylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)4NCl
CAS Number:
Molecular Weight:
109.60
MDL number:
UNSPSC Code:
12352107
EC Index Number:
200-880-8
Product 8.45105 is not currently sold in your country. Contact Technical Service

Quality Level

form

powder

potency

50 mg/kg LD50, oral (Rat)
537 mg/kg LD50, skin (Rat)

pH

6-8 (20 °C, 100 g/L in H2O)

mp

268 °C

density

1.19 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C4H12N.ClH/c1-5(2,3)4;/h1-4H3;1H/q+1;/p-1

InChI key

OKIZCWYLBDKLSU-UHFFFAOYSA-M

Analysis Note

Colour (visual): white
Appearance of substance (visual): powder
Assay (argentometric): ≥ 98.0 %
Identity (IR): passes test
Ir (Iridium): lot specific result
Ni (Nickel): lot specific result
Cr (Chromium): lot specific result
Cu (Copper): lot specific result
Fe (Iron): lot specific result
Zn (Zinc): lot specific result
Methanol (GC): lot specific result

Legal Information

MSYNTH is a trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Aquatic Chronic 2 - Skin Irrit. 2 - STOT SE 1 Oral

target_organs

Central nervous system

Storage Class

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service