8.45053
Isopropylamine
Msynth™plus
Synonym(s):
Isopropylamine, 2-Propylamin, 2-Aminopropan
About This Item
vapor pressure
631 hPa ( 20 °C)
assay
≥99.5% (GC)
autoignition temp.
330 °C
potency
550 mg/kg LD50, oral (Rat)
>400 mg/kg LD50, skin (Rabbit)
expl. lim.
2-10.4 % (v/v)
pH
13 (20 °C, 700 g/L in H2O)
kinematic viscosity
0.47 cSt(20 °C)
bp
32 °C/1013 hPa
transition temp
flash point -37 °C
density
0.69 g/cm3 at 20 °C
storage temp.
2-30°C
InChI
1S/C3H9N/c1-3(2)4/h3H,4H2,1-2H3
InChI key
JJWLVOIRVHMVIS-UHFFFAOYSA-N
Analysis Note
Colour number (Hazen): ≤ 20
Assay (GC, area%): ≥ 99.5 % (a/a)
Density (d 20 °C/ 4 °C): 0.687 - 0.688
Water (K. F.): ≤ 0.20 %
Identity (IR): passes test
Acetone: lot specific result
2-Propanol: lot specific result
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service