Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

8.43914

Sigma-Aldrich

Vinylphosphonic acid, 90%

for synthesis

Synonym(s):

Vinylphosphonic acid, 90%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H5O3P
CAS Number:
Molecular Weight:
108.03
MDL number:
UNSPSC Code:
12352106
EC Index Number:
217-123-2

Quality Level

form

liquid

potency

>2000 mg/kg LD50, oral (Rat)

pH

1.5 (30 °C, 50 g/L in H2O)

mp

36 °C

transition temp

flash point 113 °C

density

1.37 g/cm3 at 30 °C

storage temp.

2-30°C

InChI

1S/C2H5O3P/c1-2-6(3,4)5/h2H,1H2,(H2,3,4,5)/p-2

InChI key

ZTWTYVWXUKTLCP-UHFFFAOYSA-L

Analysis Note

Colour (visual): colourless to pale yellow
Appearance of substance (visual): liquid
Assay (NMR): ≥ 91.0 %
Phosphoric acid (NMR): ≤ 5.0 %
other org. and inorg. Phophor compounds (NMR): ≤ 4.0 mol%
Water (K. F.): ≤ 0.50 %
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

pictograms

Corrosion

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class

8A - Combustible, corrosive hazardous materials

wgk_germany

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service