8.43860
Methyl fufuryl disulfide
for synthesis
Synonym(s):
Methyl fufuryl disulfide
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C6H8OS2
CAS Number:
Molecular Weight:
160.26
MDL number:
UNSPSC Code:
12141912
EC Index Number:
260-773-7
assay
≥95.0% (GC)
bp
60-61 °C/0.8 hPa
transition temp
flash point 90 °C
density
1.183 g/cm3
storage temp.
2-30°C
InChI
1S/C6H8OS2/c1-8-9-5-6-3-2-4-7-6/h2-4H,5H2,1H3
InChI key
CLSLQQCDHOZMDT-UHFFFAOYSA-N
Analysis Note
Assay (GC, area%): ≥ 95.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.180 - 1.190
Identity (IR): passes test
Density (d 20 °C/ 4 °C): 1.180 - 1.190
Identity (IR): passes test
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 3 - STOT RE 2
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
194.0 °F - closed cup
flash_point_c
90 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service