Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.43302

Sigma-Aldrich

N-Carbamoyl-2-cyanoacetamide

for synthesis

Synonym(s):

N-Carbamoyl-2-cyanoacetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H5N3O2
CAS Number:
Molecular Weight:
127.10
MDL number:
UNSPSC Code:
12352111
EC Index Number:
215-906-3

Quality Level

assay

≥98.0% (HPLC)

form

powder

mp

203-207 °C

storage temp.

2-30°C

InChI

1S/C4H5N3O2/c5-2-1-3(8)7-4(6)9/h1H2,(H3,6,7,8,9)

InChI key

QJGRPCPCQQPZLZ-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 203 °C
Melting range (upper value): ≤ 207 °C
Identity (IR): passes test

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service