Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.41347

Sigma-Aldrich

4-Bromo-1-iodobenzene

for synthesis

Synonym(s):

4-Bromo-1-iodobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4BrI
CAS Number:
Molecular Weight:
282.90
MDL number:
UNSPSC Code:
12352101
EC Index Number:
209-662-7

assay

≥99.0% (GC)

form

needles

bp

251 °C/1005 hPa

mp

89-91 °C

storage temp.

2-30°C

InChI

1S/C6H4BrI/c7-5-1-3-6(8)4-2-5/h1-4H

InChI key

UCCUXODGPMAHRL-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 89 °C
Melting range (upper value): ≤ 91 °C
Identity (IR): passes test

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service