Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.41176

Sigma-Aldrich

2-Chloro-2-phenylacetophenone

for synthesis

Synonym(s):

2-Chloro-2-phenylacetophenone, Desyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H11ClO
CAS Number:
Molecular Weight:
230.69
UNSPSC Code:
12352115
EC Index Number:
207-181-7

assay

≥99.0% (HPLC)

form

powder

mp

67-68 °C

storage temp.

2-30°C

InChI

1S/C14H11ClO/c15-13(11-7-3-1-4-8-11)14(16)12-9-5-2-6-10-12/h1-10,13H

InChI key

RXDYOLRABMJTEF-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 66 °C
Melting range (upper value): ≤ 69 °C
Identity (IR): passes test

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

target_organs

Respiratory system

Storage Class

8A - Combustible, corrosive hazardous materials

wgk_germany

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service