Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

8.41028

Sigma-Aldrich

Acetonyltriphenylphosphonium chloride

for synthesis

Synonym(s):

Acetonyltriphenylphosphonium chloride, 2-Oxopropyl triphenylphosphonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H20ClOP
CAS Number:
Molecular Weight:
354.81
MDL number:
UNSPSC Code:
12352107
EC Index Number:
214-974-1

Quality Level

form

powder

reaction suitability

reaction type: C-C Bond Formation

mp

243-245 °C

storage temp.

2-30°C

InChI

1S/C21H20OP.ClH/c1-18(22)17-23(19-11-5-2-6-12-19,20-13-7-3-8-14-20)21-15-9-4-10-16-21;/h2-16H,17H2,1H3;1H/q+1;/p-1

InChI key

XAMZZEBAJZJERT-UHFFFAOYSA-M

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service