Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.40161

Sigma-Aldrich

4-Methoxybenzyltriphenylphosphonium chloride

for synthesis

Synonym(s):

4-Methoxybenzyltriphenylphosphonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H24ClOP
CAS Number:
Molecular Weight:
418.89
UNSPSC Code:
12352107

form

solid

mp

236-238 °C

storage temp.

2-30°C

InChI

1S/C26H24OP.ClH/c1-27-23-19-17-22(18-20-23)21-28(24-11-5-2-6-12-24,25-13-7-3-8-14-25)26-15-9-4-10-16-26;/h2-20H,21H2,1H3;1H/q+1;/p-1

InChI key

YQXBNCFNXOFWLR-UHFFFAOYSA-M

Analysis Note

Assay (argentometric): ≥ 97.0 %
Melting range (lower value): ≥ 230 °C
Melting range (upper value): ≤ 235 °C
Identity (IR): passes test

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service