Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.21819

Sigma-Aldrich

Phenylmercury hydroxide

for synthesis

Synonym(s):

Phenylmercury hydroxide, Hydroxyphenylmercury

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6HgO
CAS Number:
Molecular Weight:
294.70
UNSPSC Code:
12352103
EC Index Number:
202-866-7

form

powder

solubility

50 g/L

storage temp.

2-30°C

InChI

1S/C6H5.Hg.H2O/c1-2-4-6-5-3-1;;/h1-5H;;1H2

InChI key

MSYUMXXVCSLXMR-UHFFFAOYSA-N

Analysis Note

Assay (ex Hg): ≥ 94.0 %
Identity (IR): passes test

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - STOT RE 1

Storage Class

6.1B - Non-combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service