Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.21032

Sigma-Aldrich

Pinacol

for synthesis

Synonym(s):

Pinacol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H14O2
CAS Number:
Molecular Weight:
118.17
MDL number:
UNSPSC Code:
12352104
EC Index Number:
200-933-5

assay

≥99.0% (GC)

form

solid

bp

174 °C/1013 hPa

mp

41 °C

transition temp

flash point 77 °C

density

0.967 g/cm3 at 20 °C

Storage temp.

2-30°C

InChI

1S/C6H14O2/c1-5(2,7)6(3,4)8/h7-8H,1-4H3

Inchi Key

IVDFJHOHABJVEH-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

170.6 °F

flash_point_c

77 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service