Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.20963

Sigma-Aldrich

1-Pentyne

for synthesis

Synonym(s):

1-Pentyne, Propylacetylene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8
CAS Number:
Molecular Weight:
68.12
MDL number:
UNSPSC Code:
12352003
EC Index Number:
210-987-1

vapor pressure

467 hPa ( 20 °C)

assay

≥98.0% (GC)

bp

39-41 °C/1013 hPa

mp

-106 °C

transition temp

flash point -20 °C

solubility

1.05 g/L

density

0.69-0.7 g/cm3 at 20 °C

storage temp.

2-8°C

InChI

1S/C5H8/c1-3-5-4-2/h1H,4-5H2,2H3

InChI key

IBXNCJKFFQIKKY-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.690 - 0.700
Identity (IR): passes test

pictograms

Flame

signalword

Danger

hcodes

Hazard Classifications

Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

-4.0 °F

flash_point_c

-20 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service