8.20743
N-Iodosuccinimide
for synthesis
Synonym(s):
N-Iodosuccinimide, NIS
About This Item
Recommended Products
mp
201-202 °C (decomposition)
density
2.25 g/cm3
storage temp.
2-30°C
InChI
1S/C4H4INO2/c5-6-3(7)1-2-4(6)8/h1-2H2
InChI key
LQZMLBORDGWNPD-UHFFFAOYSA-N
Analysis Note
Identity (IR): passes test
signalword
Warning
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1B
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service