8.20736
o-Iodohippuric acid
for synthesis
Synonym(s):
o-Iodohippuric acid, (2-Iodobenzoyl)glycine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
2-(I)C6H4NHCH2COOH
CAS Number:
Molecular Weight:
305.07
MDL number:
UNSPSC Code:
12352106
EC Index Number:
205-693-5
assay
≥98% (acidimetric)
form
powder
mp
170-172 °C (External MSDS)
storage temp.
2-30°C
InChI
1S/C9H8INO3/c10-7-4-2-1-3-6(7)9(14)11-5-8(12)13/h1-4H,5H2,(H,11,14)(H,12,13)
InChI key
CORFWQGVBFFZHF-UHFFFAOYSA-N
Analysis Note
Assay (acidimetric): ≥ 98
Identity (IR-spectrum): passes test
Melting range: 172 - 176
o- Iodobenzoic acid (HPLC, contra external standard): ≤ 0.1 %
Identity (IR-spectrum): passes test
Melting range: 172 - 176
o- Iodobenzoic acid (HPLC, contra external standard): ≤ 0.1 %
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service