Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.20496

Sigma-Aldrich

Boron hydride dimethylamine

for synthesis

Synonym(s):

Boron hydride dimethylamine, Dimethylaminoborane, Borane-dimethylamine-complex

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NHBH3
CAS Number:
Molecular Weight:
58.92
MDL number:
UNSPSC Code:
12352402
EC Index Number:
200-823-7

vapor pressure

<5 hPa ( 20 °C)

form

solid

autoignition temp.

175 °C

potency

59 mg/kg LD50, oral (Rat)
210 mg/kg LD50, skin (Rabbit)

reaction suitability

reagent type: reductant

pH

8-9 (20 °C, 10 g/L in H2O)

mp

33-36 °C

transition temp

flash point 43 °C

solubility

120 g/L

density

0.73 g/cm3 at 20 °C

bulk density

300‑350 kg/m3

storage temp.

2-30°C

InChI

1S/C2H7N.B/c1-3-2;/h3H,1-2H3;

InChI key

RJTANRZEWTUVMA-UHFFFAOYSA-N

Analysis Note

Assay (iodometric): ≥ 97.0 %
Melting range (lower value): ≥ 33 °C
Melting range (upper value): ≤ 36 °C
Identity (IR): passes test

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Sol. 1 - Skin Sens. 1

Storage Class

4.1B - Flammable solid hazardous materials

wgk_germany

WGK 2

flash_point_c

43 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service