Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.20180

Sigma-Aldrich

Tetrabromo-p-benzoquinone

for synthesis

Synonym(s):

Tetrabromo-p-benzoquinone, p-Bromanile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
1-O=C6Br4-4-(=O)
CAS Number:
Molecular Weight:
423.68
UNSPSC Code:
12352115
EC Index Number:
207-679-4

form

powder

mp

300 °C

storage temp.

2-30°C

InChI

1S/C6Br4O2/c7-1-2(8)6(12)4(10)3(9)5(1)11

InChI key

LWHDQPLUIFIFFT-UHFFFAOYSA-N

Analysis Note

Assay (ex Br): ≥ 96.0 %
Identity (IR): passes test

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service