Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8200SCPKIT

Millipore

Large Replacement Parts for 8200 Series Stirred Cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116500
eCl@ss:
32019190

shipped in

ambient

Components

Cap assembly - Part no. 1524903 - Qty/Kit- 1
Stirrer assembly - Part no. 15256 - Qty/Kit- 1
base - Part no. 21011 - Qty/Kit- 1

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service