Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.18688

Sigma-Aldrich

Dibutyl maleate

Synonym(s):

Maleic acid dibutyl ester

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116105

form

liquid

Quality Level

signalword

Warning

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1 - STOT RE 2 Oral

target_organs

Kidney

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

248.0 °F

flash_point_c

120 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service