Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.18638

Sigma-Aldrich

N,N-Dimethylcyclohexylamine

for synthesis

Synonym(s):

N,N-Dimethylcyclohexylamine, N-Cyclohexyldimethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11N(CH3)2
CAS Number:
Molecular Weight:
127.23
MDL number:
UNSPSC Code:
12352116
EC Index Number:
202-715-5

vapor pressure

3.6 hPa ( 20 °C)

Quality Level

assay

≥99.0% (GC)

form

liquid

autoignition temp.

215 °C

potency

272 mg/kg LD50, oral (Rat)
>400 mg/kg LD50, skin (Rat)

expl. lim.

3.6-19 % (v/v)

pH

12 (20 °C, 5 g/L in H2O)

bp

160-161 °C/1013 hPa

mp

-60 °C

transition temp

flash point 41 °C

solubility

13.4 g/L

density

0.85 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C8H17N/c1-9(2)8-6-4-3-5-7-8/h8H,3-7H2,1-2H3

InChI key

SVYKKECYCPFKGB-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.848 - 0.849
Identity (IR): passes test

signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

102.2 °F - closed cup

flash_point_c

39 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service