Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.18543

Sigma-Aldrich

1-Octen-3-ol

for synthesis

Synonym(s):

1-Octen-3-ol, Vinyl pentyl carbinol, Pentyl vinyl carbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH(OH)CH=CH2
CAS Number:
Molecular Weight:
128.21
MDL number:
UNSPSC Code:
12352104
EC Index Number:
222-226-0

vapor pressure

1 hPa ( 20 °C)

assay

≥98.0% (GC)

autoignition temp.

245 °C

potency

340 mg/kg LD50, oral (Rat)
3300 mg/kg LD50, skin (Rabbit)

expl. lim.

0.9-8 % (v/v)

kinematic viscosity

5.3 cSt(23 °C)

bp

173-175 °C/1013 hPa

mp

-49 °C

transition temp

flash point 68 °C

density

0.83-0.85 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C8H16O/c1-3-5-6-7-8(9)4-2/h4,8-9H,2-3,5-7H2,1H3

InChI key

VSMOENVRRABVKN-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.836 - 0.840
Identity (IR): passes test

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service