Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.18511

Sigma-Aldrich

4-Cyanobenzyltriphenylphosphonium chloride

for synthesis

Synonym(s):

4-Cyanobenzyltriphenylphosphonium chloride, Triphenyl-4-cyanobenzylphosphonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
413.88
UNSPSC Code:
12352107

Quality Level

form

crystals

mp

326-338 °C

storage temp.

2-30°C

InChI

1S/C26H21NP.ClH/c27-20-22-16-18-23(19-17-22)21-28(24-10-4-1-5-11-24,25-12-6-2-7-13-25)26-14-8-3-9-15-26;/h1-19H,21H2;1H/q+1;/p-1

InChI key

GXMFPJIKPZDNCS-UHFFFAOYSA-M

General description

for synthesis

Application

4-Cyanobenzyltriphenylphosphonium chloride for synthesis. CAS 20430-33-5, molar mass 413.88 g/mol.

Analysis Note

Assay (argentometric)≥ 98.0 %Identity (IR)passes test

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service