Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.14702

Sigma-Aldrich

4,5-Diamino-6-hydroxy-2-pyrimidinethiol hemisulfate-1,5-hydrate

for synthesis

Synonym(s):

4,5-Diamino-6-hydroxy-2-pyrimidinethiol hemisulfate-1,5-hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14N8O6S3 · xH2O
CAS Number:
Molecular Weight:
414.44 (anhydrous basis)
UNSPSC Code:
12352105

Quality Level

assay

≥97.0% (HPLC)

form

powder

mp

235 °C

storage temp.

2-30°C

Analysis Note

Assay (HPLC, area%): ≥ 97.0 % (a/a)
Identity (IR): passes test

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service