Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.14672

Sigma-Aldrich

(-)-[(8,8-Dichlorocamphoryl)-sulfonyl]-oxaziridine

for synthesis

Synonym(s):

(-)-[(8,8-Dichlorocamphoryl)-sulfonyl]-oxaziridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13Cl2NO3S
CAS Number:
Molecular Weight:
298.19
MDL number:
UNSPSC Code:
12352119

Quality Level

form

powder

mp

180-185 °C

storage temp.

2-30°C

InChI

1S/C10H13Cl2NO3S/c1-7(2)6-3-4-8(7)5-17(14,15)13-10(8,16-13)9(6,11)12/h6H,3-5H2,1-2H3/t6-,8-,10+,13?/m1/s1

InChI key

HAMBQYFZDBYWHU-VYBWYVTGSA-N

Analysis Note

Assay (ex Cl): ≥ 97.0 %
Melting range (lower value): ≥ 180 °C
Melting range (upper value): ≤ 185 °C
Spec. rotation [α²0/D (c=0.5 in chloroform): -92.5 - -87.5 °
Identity (IR): passes test

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service