Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

8.14671

Sigma-Aldrich

N,N-Diethylhydroxylamine

(85% solution in water) for synthesis

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352116

vapor pressure

43 hPa ( 25 °C)

bp

125-130 °C/1013 hPa

transition temp

flash point 46 °C

density

0.9 g/cm3 at 20 °C

storage temp.

2-30°C

Analysis Note

Assay (HClO₄): 80.0 - 90.0 %
Density (d 20 °C/ 4 °C): 0.900 - 0.905

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service